Learn More
anti-H-2Kk, Clone: 16-3-22S, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128202
Catalog No. NBP128202
Add to cart
Description
Description
MHC Class I H-2Kk Monoclonal antibody specifically detects MHC Class I H-2Kk in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation.
Specifications
Specifications
H-2Kk
16-3-22S
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246
Mouse
IgG2a Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
An epitope on mouse H-2Kk MHC class I alloantigen
Flow Cytometry, Immunoprecipitation
0.5 mg/mL
Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10-1:500
Purified
H2-K1
C3H mouse splenocytes
Protein A or G purified
RUO
Primary
14972
Mouse