Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HAX-1, Clone: 3C5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP130169

Catalog No. NBP130169


Add to cart

Description

Description

HAX-1 Monoclonal antibody specifically detects HAX-1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

HAX-1
3C5
Unconjugated
PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Purified
HAX1
Recombinant human HAX1 (1-279aa) purified from E. coli
Protein G purified
RUO
Primary
10456
Human
ELISA, Western Blot
1.0 mg/mL
Western Blot 1:1000-1:2000, ELISA 1:100-1:2000
NP_006109
FLJ17042, FLJ93803, HAX-1, HCLS1 (and PKD2) associated protein, HCLS1 associated protein X-1, HCLS1-associated protein X-1, HCLSBP1, HS1 binding protein, HS1-associating protein X-1, HS1-binding protein 1, HS1BP1FLJ18492, HSP1BP-1, SCN3
Mouse
IgG2b Kappa
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human HAX1 protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only