Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chorionic Gonadotropin alpha Chain (hCG alpha) Antibody (SPM552), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234794AF405
Description
Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Chorionic Gonadotropin alpha Chain (HCG alpha) | |
Monoclonal | |
Alexa Fluor 405 | |
CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA | |
Mouse | |
13 kDa | |
0.1 mL | |
Primary | |
This MAb reacts with a protein of ∼13kDa, identified as alpha sub-unit of HCG. HCG is a glycoprotein, which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the ? and ? subunits. The subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
SPM552 | |
Western Blot, ELISA, Immunohistochemistry-Paraffin | |
CGA | |
Recombinant Chorionic Gonadotropin alpha Chain (hCG alpha) protein (Uniprot: P01215) | |
Protein A or G purified | |
RUO | |
1081 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction