Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-hCG alpha, Alexa Fluor 488, Clone: SPM552, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP234794AF488

Catalog No. NBP234794X


Add to cart

Description

Description

Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal antibody specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.
Specifications

Specifications

Chorionic Gonadotropin alpha Chain (HCG alpha)
SPM552
Western Blot, ELISA, Immunohistochemistry-Paraffin
Purified
CGA
Recombinant hCG alpha protein
13 kDa
0.1 ml
Store at 4C in the dark.
Monoclonal
This MAb reacts with a protein of ~13kDa, identified as alpha sub-unit of HCG. HCG is a glycoprotein, which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the ? and ? subunits. The subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH).
ELISA, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA
Mouse
IgG1 Kappa
Protein A purified
RUO
Primary
1081
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only