Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-hCG Beta, Alexa Fluor 647, Clone: SPM105, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP234392AF647

Catalog No. NBP234392Y


Add to cart

Description

Description

Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal antibody specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Chorionic Gonadotropin beta Chain (HCG beta)
SPM105
Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
CGB
Recombinant hCG beta protein
22 kDa
0.1 ml
Cancer
Primary
1082
Human
Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
CGB3choriogonadotropin subunit beta, CGB5, CGB7, CGB8, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit, chorionic gonadotropin, beta polypeptide, hCGB
Mouse
IgG1 Kappa
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
This MAb reacts with a protein of 22kDa, identified as beta sub-unit of HCG. It does not cross react with the alpha sub-unit.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only