Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HDAC3, Clone: 7G6C5, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP147529

Catalog No. NBP147529

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

HDAC3
7G6C5
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000
Ascites
HDAC3
Purified recombinant fragment of human HDAC3 (aa224-428) expressed in E. Coli.
48.8 kDa
0.1 ml
Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases
Primary
8841
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45
Mouse
IgG2a
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only