Learn More
anti-Hepatocyte Specific Antigen, Clone: EP-1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142180
Catalog No. NBP142180
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Hepatocyte Specific Antigen Monoclonal antibody specifically detects Hepatocyte Specific Antigen in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
Specifications
Hepatocyte Specific Antigen
EP-1
Unconjugated
0.1 mg/ml PBS with 0.05% Sodium Azide
Hepatocyte, HSA
The antibody reacts with the Hepatocytes antigen.
Protein A or G purified
RUO
Primary
Monoclonal antibody EP-1 stains human liver canaliculi and a subset of hepatocellular carcinomas. In frozen sections, it stains liver canaliculi strongly and may be used as a marker of this hepatic substructure. Cell preparations of hepatocellular carcinoma biopsies and cell lines are found to bind EP-1 on the cell surface. Monoclonal antibody EP-1 stains liver canaliculi in normal liver sections and is positive on the cell surface of human liver carcinomas. Monoclonal antibodies to liver cell processes are useful in the identification of hepatic carcinomas and normal organ structures. Two monoclonal antibodies to different liver substructures are offered which bind to subsets of hepatic carcinomas.
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
0.1 mg/mL
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1-10-1:2000, Immunohistochemistry-Frozen 1:10-1:500
Purified
Mouse
IgG2b
0.1 mg
Store at 4C. Do not freeze.
Monoclonal
Human