Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HGK/MAP4K4, Clone: 3C7B5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128855

Catalog No. NBP128855


Add to cart

Description

Description

HGK/MAP4K4 Monoclonal antibody specifically detects HGK/MAP4K4 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

HGK/MAP4K4
3C7B5
Western Blot 1:500-1:2000, ELISA 1:10000
Ascites
MAP4K4
Purified recombinant fragment of MAP4K4(aa400-500) expressed in E. Coli.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Specific for MAP4K4.
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
EC 2.7.11, EC 2.7.11.1, FLH21957, FLJ10410, hepatocyte progenitor kinase-like/germinal center kinase-like kinase, HGKFLJ20373, HPK/GCK-like kinase HGK, KIAA0687, MAPK/ERK kinase kinase kinase 4, MEK kinase kinase 4, MEKKK 4, mitogen-activated protein kinase kinase kinase kinase 4, Nck-interacting kinase, NIKFLJ90111
Mouse
IgG1
0.1 ml
Protein Kinase
Primary
9448
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only