Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Histone Deacetylase 4/HDAC4, Clone: 7B2, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP151594

Catalog No. NBP151594

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Histone Deacetylase 4/HDAC4
7B2
Western Blot 1:500-1:2000, ELISA 1:10000
Ascites
HDAC4
Purified recombinant fragment of human HDAC4 expressed in E. Coli.
119 kDa
0.1 ml
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases
Primary
9759
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3
Mouse
IgG1
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only