Learn More
anti-Histone H3 (Trimethyl Lys9), DyLight 550, Clone: 6F12-H4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP130141R
Catalog No. NBP130141R
Add to cart
Description
Description
Histone H3 Trimethyl Lys9 Monoclonal antibody specifically detects Histone H3 Trimethyl Lys9 in Human, Mouse, Rat, Porcine, C. elegans, Drosophila, Invertebrate, Mammal, Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Histone H3 (Trimethyl Lys9)
6F12-H4
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
P84243
H3 histone, family 3A, H3.3AH3F3H3F3B, H3.3B, histone H3.3, MGC87782, MGC87783
Mouse
IgG1 Kappa
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
All mammals including human, mouse and rat, Drosophila, C. elegans, yeast. Invertebrate/Blattella germanica (German cockroach) reactivity reported in scientific literature (PMID: 23872316).
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Purified
H3F3A
A synthetic peptide made to an N-terminal region of Histone H3 (between amino acids 1-50). [UniProt# P84243]
15 kDa
0.1 ml
Chromatin Research, Epigenetics, Histones and Modified Histones
Primary
126961
C. elegans, Drosophila, Human, Invertebrate, Mammalia, Mouse, Porcine, Rat, Yeast