Learn More
anti-Histone H4 (Monomethyl Lys20), Alexa Fluor 700, Clone: 5E10-D8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP130091AF700
Catalog No. NBP130091Z
Add to cart
Description
Description
Histone H4 Monomethyl Lys20 Monoclonal antibody specifically detects Histone H4 Monomethyl Lys20 in Human, Mouse, Rat, C. elegans, Drosophila, Mammal, Yeast samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Histone H4 (Monomethyl Lys20)
5E10-D8
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
P62805
H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Mouse
IgG1 Kappa
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
All mammals including human, mouse and rat, Drosophila, C. elegans, yeast.
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Purified
HIST4H4
Branched peptide found between amino acids 10-100 of Histone H4. [UniProt# P62805]
11 kDa
0.1 ml
Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones
Primary
121504
Human, Mouse, Rat, C. elegans, Drosophila, Mammalia, Yeast