Learn More
anti-HPV16, Clone: 716-325, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11017341
Catalog No. NB11017341
Add to cart
Description
Description
HPV16 E7 Monoclonal specifically detects HPV16 E7 in Virus samples. It is validated for Western Blot, Sandwich ELISA.
Specifications
Specifications
HPV16
716-325
Western Blot 1:500-1:1000, Sandwich ELISA 1:100-1:2000
Purified
Mouse
IgG2a
0.2 mg
Virology Bacteria and Parasites
Primary
HPV oncoprotein E7, types 16 & 18 (both monomer and dimer forms according WB)
Western Blot, ELISA
Unconjugated
PBS (pH 7.2) with 0.09% Sodium Azide
Human Papilloma Virus Type 16
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with HPV oncoprotein E7, type 16.
Protein G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Virus