Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HSP70/HSPA1A, DyLight 350, Clone: 2A4, Novus Biologicals
Manufacturer: Novus Biologicals NBP121693UV
Specifications
HSP70/HSPA1A | |
2A4 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
P08107 | |
dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
Mouse | |
IgM | |
0.1 ml | |
Cancer, Cellular Markers, Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific | |
Primary | |
3303 | |
Human |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Purified | |
HSPA1A | |
Human recombinant HSP70 overexpressed in E.coli | |
PEG purified | |
RUO | |
Store at 4C in the dark. | |
Monoclonal | |
Detects approx 70kDa. May detect HSP70, HSC70, and HSP72. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only