Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Hsp70 interacting protein HIP, Clone: 1F1, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP126368

Catalog No. NBP126368

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Hsp70 interacting protein HIP
1F1
Unconjugated
PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide
Purified
ST13
Recombinant human HIP/ST13 (1-369aa) purified from E. coli
Protein G purified
RUO
Primary
6767
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 1:1000-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:25-50
NP_003923
AAG2, aging-associated protein 2, FAM10A1FAM10A4, heat shock 70kD protein binding protein, Hip, HIPFLJ27260, HOP, hsc70-interacting protein, Hsp70-interacting protein, HSPABP1, P48MGC129952, PRO0786, Progesterone receptor-associated p48 protein, Protein FAM10A1, Putative tumor suppressor ST13, Renal carcinoma antigen NY-REN-33, SNC6HSPABP, suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein), suppression of tumorigenicity 13 (colon carcinoma) (Hsp70-interacting protein), Suppression of tumorigenicity 13 protein
Mouse
IgG1 Kappa
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only