Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgM Antibody (P5E2), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP178603
Description
Human IgM Monoclonal specifically detects Human IgM in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Radioimmunoassay.Specifications
Human IgM | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100 - 1:400, Flow Cytometry 1 ug/ml, ELISA 1 ug/ml, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
P5E2 | |
Unconjugated | |
AGM1, MU, VH | |
Purified Human Serum IgM | |
100 μg | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction