Learn More
anti-IFN-gamma/Granzyme B, DyLight 755, Clone: CLB-GB11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP150071IR
Catalog No. NBP150071IR
Add to cart
Description
Description
Granzyme B Monoclonal antibody specifically detects Granzyme B in Human, Primate samples. It is validated for Flow Cytometry.
Specifications
Specifications
Granzyme B
CLB-GB11
Flow Cytometry
Purified
GZMB
Human NK cell line YT-INDY-derived granzyme B
Protein A purified
RUO
Primary
3002
Human, Primate
Flow Cytometry
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GZMB, HLP, SECT
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
The mouse monoclonal antibody CLB-GB11 recognizes granzyme B, a 31 kDa serine protease expressed in activated Tc cells and NK cells.