Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse IgA, Isotype Control, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197030

Catalog No. NBP197030


Add to cart

Description

Description

IgA Monoclonal antibody specifically detects IgA in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications

Specifications

IgA
Unconjugated
0.02 M Potassium Phosphate, 0.5 M Sodium Chloride, pH 7.2. Stabilizer: None. with 0.01% Sodium Azide
Immunoglobulin A
Mouse
Size Exclusion Chromatography
RUO
Isotype Control
Monoclonal
In an Ouchterlony double diffusion assay the material is non-reactive with antisera to mouse IgG1, IgG2a, IgG2b, IgG3 and IgM. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse IgA alpha and anti-Mouse serum.
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Western Blot 1:100 - 1:2000, Flow Cytometry 1:1000 - 1:5000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Concentrated
IGHA1
IgA
1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Primary
3493
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only