Learn More
anti-IGFBP-1, Clone: A8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11068179
Catalog No. NB11068179
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IGFBP-1 Monoclonal antibody specifically detects IGFBP-1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
IGFBP-1
A8
Western Blot 1:100-1:2000, ELISA indirect
Purified
IGFBP1
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified IGFBP-1 from human amniotic fluid.
Protein G purified
RUO
Primary
3484
Human
Western Blot, ELISA
Unconjugated
PBS (pH 7.4) with 0.09% Sodium Azide
alpha-pregnancy-associated endometrial globulin, amniotic fluid binding protein, binding protein-25, binding protein-26, binding protein-28, growth hormone independent-binding protein, hIGFBP-1, IBP1, IBP-1, IGF-binding protein 1, IGFBP-1, IGF-BP25, insulin-like growth factor binding protein 1, insulin-like growth factor-binding protein 1, Placental protein 12, PP12AFBP
Mouse
IgG1
0.2 mg
Store at 4C. Do not freeze.
Monoclonal
Human IGFBP1