Learn More
Rockland Immunochemicals Anti-MOUSE IgG (H&L) (GOAT) Antibody Peroxidase Conjugated - 610-1302
SDP
Supplier:  Rockland Immunochemicals 6101302
Encompass_Preferred
Anti-Mouse secondary antibody conjugated to horseradish peroxidase (HRP) generated in goat detects specifically Mouse IgG whole molecule. This anti-Mouse HRP antibody is suitable for ELISA, Sandwich ELISA, titration assays, western-blot, immunoprecipitation, Immunohistochemistry as well as other HRP antibody based assays. Specific conditions for reactivity and signal detection should be optimized by the end user.
Catalog No. NC0332513
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.