Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse IgG, AP, Isotype Control, Novus Biologicals™

Mouse Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197220

Catalog No. NBP197220


Add to cart

Description

Description

IgG antibody specifically detects IgG in Mouse samples. It is validated for SDS-Page.
Specifications

Specifications

IgG
AP
0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, containing 10 mg/ml BSA (Immunoglobulin and Protease free) with 0.1% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
Mouse
Multi-step
RUO
Isotype Control
3500
Mouse
SDS-Page
SDS-Page 1:100 - 1:2000
Conjugated
IGHG1
IgG
1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Alkaline Phosphatase (calf intestine), anti-Mouse IgG and anti-Mouse Serum.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only