Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Antibody (ICO-97), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234647APC
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry.Specifications
IgG | |
Monoclonal | |
APC | |
Immunoglobulin G, ImmunoglobulinG | |
Mouse | |
75 kDa | |
0.1 mL | |
Immunology | |
3500 | |
Human | |
Purified |
Flow Cytometry | |
ICO-97 | |
Flow Cytometry | |
IGHG1 | |
Purified human Ig Gamma Chain | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This monoclonal antibody is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant. | |
Store at 4C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction