Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse IgG, Biotin, Isotype Control, Novus Biologicals™

Mouse Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197078

Catalog No. NBP197078


Add to cart

Description

Description

IgG antibody specifically detects IgG in Mouse samples. It is validated for SDS-Page.
Specifications

Specifications

IgG
Biotin
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with No Preservative
immunoglobulin heavy constant gamma 1 (G1m marker)
Mouse
Ion exchange chromatography
RUO
Isotype Control
3500
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
SDS-Page
SDS-Page 1:100 - 1:2000
Lyophilized
IGHG1
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Mouse IgG and anti-Mouse Serum.
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only