Learn More
IgG Antibody (ICO-97), DyLight 650, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP234647C
Catalog No. NBP234647C
Add to Cart
Description
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
IgG
Monoclonal
DyLight 650
Immunoglobulin G, ImmunoglobulinG
Mouse
75 kDa
0.1 mL
Immunology
3500
Human
Purified
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
ICO-97
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
IGHG1
Purified human Ig Gamma Chain
Protein A or G purified
RUO
Primary
Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This monoclonal antibody is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Store at 4C in the dark.
IgG1 κ
Product Suggestions
Product Suggestions