Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MOUSE IGG (H+L) AB HSA  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52100185
Affinity purified polyclonal antibody to Mouse IgG, both heavy and light chains (whole IgG) made in Goat. Product has been cross-adsorbed with human serum to reduce cross-reactivity to human immunoglobulin.
Catalog No. 50-668-26