Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody, GE Healthcare: PROMO

Product

Manufacturer:  GE Healthcare Life Sciences NA931100ULPM

Catalog No. 45-001-275LR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Host Species: Sheep
Isotype: IgG
Species Reactivity: Ms

 

Specifications

Specifications

IgG, Whole Molecule
Affinity Purified
IgG
Secondary
HRP
Sheep
0.1mL
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit