Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse IgG1, kappa L Chain, DyLight 488, Clone: P3.6.2.8.1 Isotype Control, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP143319G

Catalog No. NBP143319G

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

IgG1 Kappa Light Chain
P3.6.2.8.1
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500
Liquid
IgG1 Kappa
RUO
Isotype Control
Monoclonal
Flow Cytometry, Immunoprecipitation, Western Blot
DyLight 488
50 mM Sodium Borate. with 0.05% Sodium Azide
Mouse
500 Tests
Store at 4C in the dark.
Primary
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only