Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat anti-Mouse IgG2a (Heavy chain) Secondary Antibody, Biotin, Invitrogen™
Rat Monoclonal Secondary Antibody
Supplier: Thermo Scientific MA516788
Description
Rat anti Mouse IgG2a:Biotin, clone LO-MG2a-7 recognizes the gamma 2a heavy chain of mouse immunoglobulin and does not cross-react with other murine immunoglobulin classes or subclasses.
Mouse IgG2a (Heavy chain) Monoclonal antibody specifically detects Mouse IgG2a (Heavy chain) in Mouse samples. It is validated for ELISA.Specifications
Mouse IgG2a (Heavy chain) | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide | |
Purified IgG from BALB/c mice | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
ELISA | |
LO-MG2a-7 | |
Biotin | |
Rat | |
Affinity chromatography | |
RUO | |
Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction