Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse IgG2a, κ, BV711 Isotype Control, Clone: G155-178, BD

Manufacturer:  BD Biosciences 563345

Catalog No. BDB563345


Add to cart

Description

Description

Host Species: Mouse
Conjugate: BV711
Clone: G155-178
Isotype: IgG2a, kappa
Immunogen: TNP-keyhole limpet hemocyanin

Isotype Control

Specifications

Specifications

Flow Cytometry
0.2mg/mL
Affinity Purified
TNP-keyhole limpet hemocyanin
50μg
Primary
G155-178
Brilliant Violet 711
Murine
IgG2a, kappa
RUO
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit