Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse IgG2b, Isotype Control, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196979

Catalog No. NBP196979


Add to cart

Description

Description

IgG2b Lambda Monoclonal antibody specifically detects IgG2b Lambda in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications

Specifications

IgG2b Lambda
M2BL
Western Blot 1:100 - 1:2000, Flow Cytometry 1:1000 - 1:5000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Concentrated
IgG2b
1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Primary
Mouse
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.5 M Sodium Chloride, pH 7.2. Stabilizer: None. with 0.01% Sodium Azide
Mouse
Protein A purified
RUO
Isotype Control
In an Ouchterlony double diffusion assay the material is non-reactive with antisera to mouse IgG1, IgG2a, IgG3, IgM and IgA. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse IgG and anti-Mouse serum.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only