Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG2b Kappa Isotype Control (M2BK), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP196980
Description
IgG2b Kappa Monoclonal specifically detects IgG2b Kappa in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.Specifications
IgG2b Kappa | |
M2BK | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:1000 - 1:5000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Fluorophore-linked immunosorbent assay 1:100 - 1:2000 | |
Mouse | |
1 mg | |
Isotype Control (Primary) | |
Mouse | |
Concentrated |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.5 M Sodium Chloride, pH 7.2. Stabilizer: None. with 0.01% Sodium Azide | |
Protein A purified | |
RUO | |
Isotyping assay resulted non-reactive with antisera to mouse IgG1, IgG2a, IgG3, IgA, IgM | |
Store at 4C. Do not freeze. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction