Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse IgG3, κ Isotype Control, Clone: J606, BD

Manufacturer:  BD Biosciences 563627

Catalog No. BDB563627


Add to cart

Description

Description

Host Species: Mouse
Clone: J606
Isotype: IgG3, kappa

Isotype Control

Specifications

Specifications

Control, Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Mouse
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with APC-H7 under optimum conditions, and unconjugated antibody and APC-H7 were removed.
RUO
Isotype Control
Monoclonal
J606
APC-H7
Affinity Purified
Mouse BALB/c IgG3, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype Control
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit