Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse IgM, Isotype Control, Novus Biologicals™

Mouse Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197007

Catalog No. NBP197007


Add to cart

Description

Description

IgM antibody specifically detects IgM in Mouse samples. It is validated for Western Blot, ELISA, SDS-Page.
Specifications

Specifications

IgM
Unconjugated
0.1 M Tris Chloride, 0.5 M Sodium Chloride, pH 8.0. Stabilizer: None. with 0.1% Sodium Azide
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
Mouse
Size Exclusion Chromatography
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse Serum and anti-Mouse IgM (mu chain specific). No reaction was observed against anti-Mouse IgG F(c). Some light chain cross-reactivity will occur with anti-Mouse IgG.
Western Blot, ELISA, SDS-Page
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, SDS-Page 1:100 - 1:2000
Concentrated
IGHM
IgM
1 mg
Immunology
Isotype Control
3507
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only