Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MOUSE IGM (MU) AB HSA CY5  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300375
Affinity purified polyclonal antibody to Mouse IgM heavy chains (mu) made in Goat and labeled with Cy5. Cross-reactivity with human serum has been minimized with affinity procedures. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-670-65