Learn More
anti-IKK beta, Clone: 10A9B6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233214
Catalog No. NBP233214
Add to cart
Description
Description
IKK beta Monoclonal antibody specifically detects IKK beta in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Gel Super Shift Assays, Flow (Intracellular).
Specifications
Specifications
IKK beta
10A9B6
Unconjugated
PBS - BSA free with No Preservative
Purified
IKBKB
Full-length human IKKb/IKK2 recombinant protein was used as immunogen (NP_001547).
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
An approx. 87 kDa band should be observed.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
0.5 mg/mL
Western Blot 2-5 ug/ml, Simple Western 1:12.5, Flow Cytometry 1ul/1 million cells, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2-5 ug/10^6 cells, Gel Super Shift Assays, Flow (Intracellular)
O14920
EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta
Mouse
IgG1 Kappa
0.1 mg
Cancer, Immunology, Innate Immunity, mTOR Pathway, Phospho Specific, Protein Kinase
Primary
3551
Human, Mouse