Learn More
anti-IL-17B, DyLight 350, Clone: MM0373-3D62, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211672UV
Catalog No. NBP211672UV
Add to cart
Description
Description
IL-17B Monoclonal antibody specifically detects IL-17B in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
IL-17B
MM0373-3D62
Western Blot, Flow Cytometry
Q9UHF5
Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20
Mouse
IgG1
0.1 ml
Cytokine Research
Primary
27190
Flow Cytometry, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Purified
IL17B
Produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-17B
>95% pure by SDS-PAGE
RUO
Store at 4C in the dark.
Monoclonal
Human