Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-7R alpha/CD127 Mouse anti-Human, DyLight 550, Clone: 22F3D9, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222376R
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-7R alpha/CD127 | |
Monoclonal | |
DyLight 550 | |
NP_002176.2 | |
IL7R | |
Amino acids 141-237 of human CD127 were used as the immunogen for the antibody. | |
0.1 mL | |
Apoptosis, Cytokine Research, Immunology, Innate Immunity | |
3575 | |
Store at 4C in the dark. | |
IgG2b λ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
22F3D9 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only