Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Insulin, APC, Clone: 2D11-H5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233146APC

Catalog No. NBP233146J


Add to cart

Description

Description

Insulin Monoclonal antibody specifically detects Insulin in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

Insulin
2D11-H5
Flow Cytometry
Purified
INS
The antibody reacts with the Insulin antigen.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Insulin is a protein consisting of an alpha-chain of 21 amino acids and a beta-chain of 30 amino acids and produced in the beta-cells of the pancreas. 2D11-H5 is a specific insulin mAb as tested by ELISA and on human pancreatic tissue. Source: A BALB/c mouse was immunised with swine insulin. The mouse spleen cells were fused with SP2/0 Ag14 cells and the clones were selected on anti-insulin activity.
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Mouse
IgG1
0.1 ml
Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers
Primary
3630
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only