Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Insulin, Clone: ISL-8J, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141345

Catalog No. NBP141345


Add to cart

Description

Description

Insulin Monoclonal antibody specifically detects Insulin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Insulin
ISL-8J
Unconjugated
Lyophilized from 1.2% sodium acetate, with 2mg BSA and 0.01mg NaN3 as preservative. with No Preservative
Purified
INS
The immunogen for this antibody is produced against Insulin.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with deionized water.
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
LYOPH
Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 0.4-1 ug/ml
P01308
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Mouse
IgG1
0.1 mg
Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers
Primary
3630
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only