Learn More
anti-Integrin alpha 2b/CD41, Clone: CLB-48, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP142310
Catalog No. NBP142310
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Integrin alpha 2b/CD41
CLB-48
Immunocytochemistry/Immunofluorescence 1:10-1:2000
P08514
CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK
Mouse
IgG1
0.2 ml
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers
Primary
3674
Immunocytochemistry, Immunofluorescence
Unconjugated
20mM Tris (pH 8.0), 0.15M NaCl, 10 mg BSA and 0.001% Merthiolate with No Preservative
Ion Exchange Chromatography
ITGA2B
The antibody reacts with human trombocytes and megakaryocytes.
Ion exchange chromatography
RUO
Store at 4C. Do not freeze.
Monoclonal
Human