Learn More
anti-Integrin alpha 6/CD49f, Clone: CLB-701, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142480
Catalog No. NBP142480
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Integrin alpha 6/CD49f Monoclonal antibody specifically detects Integrin alpha 6/CD49f in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
Specifications
Integrin alpha 6/CD49f
CLB-701
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Ion Exchange Chromatography
ITGA6
The antibody reacts with the CD49f antigen.
Ion exchange chromatography
RUO
Store at 4C. Do not freeze.
Monoclonal
The monoclonal antibody is directed against the CD49f antigen, molecular weight 30/31, 120 kD (GP Ic or VLA-6 alpha-chain), which can form distinct complexes with either the CD29 antigen (GP IIa or VLA beta chain), resulting in the VLA-6 (alpha-6, beta-1) complex which is expressed on human platelets, or with the beta-4 integrin resulting in the alpha-6, beta-4 complex which is expressed on various human epithelial cells. Platelet VLA-6 functions as a laminin receptor and platelet adhesion to laminin is inhibited by this antibody. The monoclonal antibody reacts with platelets, megakaryocytes, T-lymphocytes and common acute lymphoblastic leukemia cells (alpha-6, beta-1). In immunohistology the monoclonal antibody reacts with epithelial cells of a variety of tissues, peripheral nerves, microvascular endothelial cells, placenta cyto- and syncytotrophoblasts.
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
20mM Tris (pH 8.0), 0.15M NaCl, 10 mg BSA and 0.001% Merthiolate with No Preservative
CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6
Mouse
IgG2a
0.2 ml
Signal Transduction
Primary
3655
Human