Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Integrin beta 1/CD29, Clone: DF7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142318

Catalog No. NBP142318


Add to cart

Description

Description

Integrin beta 1/CD29 Monoclonal antibody specifically detects Integrin beta 1/CD29 in Human, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Integrin beta 1/CD29
DF7
Unconjugated
PBS solution containing 1.0% w/v BSA with 0.1% Sodium Azide
CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA
Mouse
IgG1
0.05 mg
Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary
3688
Human, Rat
Western Blot
0.1 mg/mL
Western Blot 1:100-1:2000
Purified
ITGB1
The antibody reacts with the b1 subunit of the integrin protein family.
Protein A or G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
The antibody reacts with the b1 subunit of the integrin protein family. The monoclonal antibody reacts with an extracellular epitope of the b1 integrin molecule. b1 -Integrins mediate cell-extracellular matrix interactions and share a common b-subunit. Monoclonal antibodies against this b1 -Integrin subunit are applicable in paraffin-embedded and frozen sections, immunoblotting and immunoprecipitation. Numerous studies have shown that b1 -Integrins have a distinct and wide cellular distribution and that there are alterations in malignant cells in their b1-integrin expression. Monoclonal antibody to b1 integrin is derived from the hybridoma produced by fusion between myeloma cells and Balb/c spleen cells. Wheat germ agglutinin binding glycoproteins of human fibroblasts were used as immunogen.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only