Learn More
anti-Kappa Light Chain, Clone: 4G7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1107084
Catalog No. NB1107084
Add to cart
Description
Description
Kappa Light Chain Monoclonal specifically detects Kappa Light Chain in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Kappa Light Chain
4G7
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
IGKC
Mixture of light chains of human immunoglobulin.
Protein A purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Specific for independent constantly expressed epitope of kappa chain. Reacts with free and heavy chain-associated kappa chains.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
PBS (pH 7.4) with 0.09% Sodium Azide
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809
Mouse
IgG2a
0.2 mg
Immunology
Primary
3514
Human