Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNT2 Mouse anti-Human, Mouse, Rat, Clone: S11-33, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147606

Catalog No. NBP147606


Add to cart

Description

Description

KCNT2 Monoclonal antibody specifically detects KCNT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

KCNT2
S11-33
Unconjugated
PBS (pH 7.4) and 50% Glycerol with 0.09% Sodium Azide
Purified
KCNT2
Fusion protein amino acids 564-624 of mouse Slo2.1 (Slick)
Protein G purified
RUO
Primary
343450
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
Western Blot 1-10 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 0.1-1.0 ug/ml
Q6UVM3
KCa4.2, MGC119610, MGC119611, MGC119612, potassium channel subfamily T member 2, potassium channel, subfamily T, member 2, Sequence like an intermediate conductance potassium channel subunit, SLICKMGC119613, SLO2.1, sodium activated potassium channel, sodium and chloride activated ATP sensitive potassium channel, sodium- and chloride-activated ATP-sensitive potassium channel, Sodium and chloride-activated ATP-sensitive potassium channel Slo2.1, sodium-and chloride-activated ATP-sensitive potassium channel (SLICK)
Mouse
IgG1
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Detects approx 140kDa.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only