Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Kv3.4, Clone: S72-16, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147609

Catalog No. NBP147609


Add to cart

Description

Description

Kv3.4 Monoclonal antibody specifically detects Kv3.4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Kv3.4
S72-16
Unconjugated
PBS (pH 7.4) and 50% Glycerol with 0.09% Sodium Azide
Purified
KCNC4
Synthetic peptide amino acids 175-192 of rat Kv3.4
Protein G purified
RUO
Primary
3749
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1 mg/mL
Western Blot 1-10 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 0.1-1.0 ug/ml
Q63734
HKSHIIIC, K+ channel subunit, KSHIIIC, Kv3.4, MGC126818, potassium voltage-gated channel subfamily C member 4, potassium voltage-gated channel, Shaw-related subfamily, member 4, Voltage-gated potassium channel subunit Kv3.4
Mouse
IgG1
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Detects approx 70kDa (100kDa in brain due to glycosylation).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only