Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv7.4 Antibody (S43/6), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212899AF488
Description
Kv7.4 Monoclonal specifically detects Kv7.4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Kv7.4 | |
Monoclonal | |
Alexa Fluor 488 | |
DFNA2, DFNA2A, KQT-like 4, KV7.4, potassium channel KQT-like 4, Potassium channel subunit alpha KvLQT4, potassium voltage-gated channel subfamily KQT member 4, potassium voltage-gated channel, KQT-like subfamily, member 4, Voltage-gated potassium channel subunit Kv7.4 | |
Mouse | |
Protein G purified | |
RUO | |
9132 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
S43/6 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
KCNQ4 | |
Fusion protein amino acids 2-77 of human KCNQ4 | |
0.1 mL | |
Primary | |
Detects approx 77kDa. | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction