Learn More
anti-L-Selectin/CD62L, Alexa Fluor 647, Clone: DREG56, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142795AF647
Catalog No. NBP142795Y
Add to cart
Description
Description
L-Selectin/CD62L Monoclonal antibody specifically detects L-Selectin/CD62L in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
L-Selectin/CD62L
DREG56
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
SELL
PMA-activated human peripheral blood leukocytes
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
The mouse monoclonal antibody DREG56 recognizes CD62L L-selectin, a 65-76 kDa cell surface protein, expressed by neutrophils, monocytes, and subsets of T, B, and NK cells, that interacts with specific carbohydrates exposed on activated endothelial cells. HLDA V; WS Code S056
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L
Mouse
IgG1
0.1 ml
Adaptive Immunity, Immunology, Myeloid derived Suppressor Cell, Signal Transduction
Primary
6402
Human