Learn More
anti-Lambda light chain, Alexa Fluor 488, Clone: 4C2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP145071AF488
Catalog No. NBP145071X
Add to cart
Description
Description
Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Lambda Light Chain
4C2
Flow Cytometry, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
IGLC1
The antibody 4C2 reacts with lambda light chains (22.5 kDa) of human immunoglobulin.
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
The antibody 4C2 reacts with lambda light chains (22.5 kDa) of human immunoglobulin. Cross reacts with Human. Does not cross react with Hamster, Goat, Guinea Pig, Rabbit and Sheep.
Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
IGLC, immunoglobulin lambda constant group
Mouse
IgG1
0.1 ml
Adaptive Immunity, Immunology
Primary
3536
Human