Learn More
anti-Lambda light chain, Alexa Fluor 647, Clone: LcN-2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233309AF647
Catalog No. NBP233309Y
Add to cart
Description
Description
Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Lambda Light Chain
HP6054
Western Blot 1:100-1:2000, Flow Cytometry, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen
Purified
IGLC1
Human IgG was used as immunogen to generate the Lambda LcN-2 antibody.
Protein G purified
RUO
Primary
3536
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
IGLC, immunoglobulin lambda constant group
Mouse
IgG2a Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human