Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamin A + C R453W Antibody (12A-2F5), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP177401AF405
Description
Lamin A + C R453W Monoclonal specifically detects Lamin A + C R453W in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Lamin A + C R453W | |
Monoclonal | |
Alexa Fluor 405 | |
NP_733821.1 | |
LMNA | |
Synthetic peptide corresponding to amino acids 449-457 of human Lamin A + C [Accession#: NP_733821.1] | |
0.1 mL | |
Cancer, Chromatin Modifiers, Chromatin Research, DNA replication Transcription Translation and Splicing, Nuclear Envelope Markers, Signal Transduction | |
4000 | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
12A-2F5 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
70 kDa Lamin, CDCD1, CDDC, Charcot Marie Tooth Disease Axonal Type 2B1, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LAMIN A, Lamin A/C, Lamin A/C R453W, LAMIN C, LDP1, LFP, LGMD1B, LMN 1, LMN A, LMN C, LMN1, LMNA, LMNC, PRELAMIN A, PRO1, Progeria 1 (Hutchinson Gilford Type) | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Disease-associated point-mutant R453W of Lamin A + C | |
Store at 4C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction