Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Lamin A + C R453W, Biotin, Clone: 12A-2F5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP177401B

Catalog No. NBP177401B


Add to cart

Description

Description

Lamin A + C R453W Monoclonal antibody specifically detects Lamin A + C R453W in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Lamin A + C R453W
12A-2F5
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
NP_733821.1
70 kDa Lamin, CDCD1, CDDC, Charcot Marie Tooth Disease Axonal Type 2B1, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LAMIN A, Lamin A/C, Lamin A/C R453W, LAMIN C, LDP1, LFP, LGMD1B, LMN 1, LMN A, LMN C, LMN1, LMNA, LMNC, PRELAMIN A, PRO1, Progeria 1 (Hutchinson Gilford Type)
Mouse
IgG1 Kappa
0.1 ml
Cancer, Chromatin Modifiers, Chromatin Research, DNA replication Transcription Translation and Splicing, Nuclear Envelope Markers, Signal Transduction
Primary
4000
Human
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Biotin
PBS with 0.05% Sodium Azide
Purified
LMNA
Synthetic peptide corresponding to amino acids 449-457 of human Lamin A + C [Accession#: NP_733821.1]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Disease-associated point-mutant R453W of Lamin A + C
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only